Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- VTI1B
- Entrez Gene ID
-
839208
- Gene Name
- Vesicle transport v-SNARE family protein
- Gene Aliases
-
AT1G26670, ATVTI12, T24P13.5, T24P13_5, VESICAL TRANSPORT V-SNARE 12, VTI12
- Chromosome Location
-
Chr.1: 9215770 - 9218010 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_102430.5 |
NP_564255.1 |
4 - 5 |
852 |
65 |
View Full Product Details