Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GSTU13
Entrez Gene ID
839602
Gene Name
glutathione S-transferase tau 13
Gene Aliases
AT1G27130, ATGSTU13, GLUTATHIONE S-TRANSFERASE 12, GST12, T7N9.190, T7N9_190, glutathione S-transferase tau 13
Chromosome Location
Chr.1: 9425280 - 9426897 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_102475.3 NP_174033.1 1 - 2 626 60
GenBank mRNA AF288193.1 - 1 - 2 324 60
AY044324.1 - 1 - 2 324 60
AY050343.1 - 1 - 2 372 60
AY094051.1 - 1 - 2 324 60

View Full Product Details