Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAP65-8
Entrez Gene ID
839685
Gene Name
microtubule-associated protein 65-8
Gene Aliases
AT1G27920, F13K9.3, F13K9_3, microtubule-associated protein 65-8
Chromosome Location
Chr.1: 9726655 - 9729848 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332791.1 NP_001322317.1 2 - 3 440 70
NM_102557.3 NP_174113.2 2 - 3 733 70

View Full Product Details