Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RALFL4
Entrez Gene ID
839721
Gene Name
ralf-like 4
Gene Aliases
AT1G28270, AtRALF4, F3H9.8, F3H9_8, ralf-like 4
Chromosome Location
Chr.1: 9882951 - 9884189 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_102592.3 NP_174148.1 1 - 1 735 166
GenBank mRNA AK228573.1 - 1 - 1 648 166
BT005747.1 - 1 - 1 648 166

View Full Product Details