Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- DEG15
- Entrez Gene ID
-
839726
- Gene Name
- protease-like protein
- Gene Aliases
-
AT1G28320, AtDEG15, F3H9.3, F3H9_3, degradation of periplasmic proteins 15
- Chromosome Location
-
Chr.1: 9920219 - 9924488 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_102597.5 |
NP_174153.2 |
5 - 6 |
547 |
136 |
View Full Product Details