Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SRD2
Entrez Gene ID
839756
Gene Name
snRNA activating complex family protein
Gene Aliases
AT1G28560, F3M18.1, SHOOT REDIFFERENTIATION DEFECTIVE 2
Chromosome Location
Chr.1: 10038029 - 10041544 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001123901.2 NP_001117373.2 12 - 13 1443 75
NM_001332819.1 NP_001321583.1 10 - 11 1493 75
NM_102624.4 NP_174178.2 11 - 12 1410 75
GenBank mRNA AY099624.1 - 11 - 12 1321 75
BT003404.1 - 8 - 9 853 75

View Full Product Details