Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ERD2
Entrez Gene ID
839807
Gene Name
ER lumen protein retaining receptor family protein
Gene Aliases
AERD2, ARABIDOPSIS ENDOPLASMIC RETICULUM RETENTION DEFECTIVE 2, ARABIDOPSIS THALIANA ENDOPLASMIC RETICULUM RETENTION DEFECTIVE 2, AT1G29330, ATERD2, ENDOPLASMIC RETICULUM RETENTION DEFECTIVE 2, F28N24.1
Chromosome Location
Chr.1: 10258248 - 10261232 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332846.1 NP_001321392.1 2 - 3 512 96
NM_102673.6 NP_564326.1 2 - 3 682 96
GenBank mRNA AY093240.1 - 2 - 3 490 96
BT009655.1 - 2 - 3 356 96

View Full Product Details