Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PIN1
Entrez Gene ID
843693
Gene Name
Auxin efflux carrier family protein
Gene Aliases
ARABIDOPSIS THALIANA PIN-FORMED 1, AT1G73590, ATPIN1, F6D5.2, F6D5_2, PIN-FORMED 1
Chromosome Location
Chr.1: 27659673 - 27663253 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_106017.4 NP_177500.1 1 - 2 1340 92
GenBank mRNA AF372950.1 - 1 - 2 1340 92
AY093960.1 - 1 - 2 1241 92

View Full Product Details