Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MPK15
Entrez Gene ID
843702
Gene Name
MAP kinase 15
Gene Aliases
AT1G73670, ATMPK15, F25P22.9, F25P22_9, MAP kinase 15
Chromosome Location
Chr.1: 27699871 - 27703449 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_106026.3 NP_565070.2 9 - 10 1782 65

View Full Product Details