Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPPA
Entrez Gene ID
843737
Gene Name
signal peptide peptidase
Gene Aliases
AT1G73990, F2P9.14, F2P9_14, SPPA1, signal peptide peptidase
Chromosome Location
Chr.1: 27824355 - 27829039 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001334622.1 NP_001322608.1 5 - 6 1085 90
NM_106058.3 NP_565077.2 5 - 6 1103 90

View Full Product Details