Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PUR4
Entrez Gene ID
843766
Gene Name
purine biosynthesis 4
Gene Aliases
AT1G74260, F1O17.7, F1O17_7, purine biosynthesis 4
Chromosome Location
Chr.1: 27922725 - 27927962 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_106086.5 NP_177566.3 3 - 4 1225 138

View Full Product Details