Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SFP1
Entrez Gene ID
832794
Gene Name
Major facilitator superfamily protein
Gene Aliases
AT5G27350, F21A20.60, F21A20_60
Chromosome Location
Chr.5: 9648508 - 9654426 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_122617.2 NP_568493.1 6 - 7 944 98

View Full Product Details