Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- GSH2
- Entrez Gene ID
-
832797
- Gene Name
- glutathione synthetase 2
- Gene Aliases
-
AT5G27380, AtGSH2, F21A20.90, F21A20_90, GLUTATHIONE SYNTHETASE, GSHB, glutathione synthetase 2
- Chromosome Location
-
Chr.5: 9667912 - 9671015 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_122620.4 |
NP_568495.2 |
4 - 5 |
783 |
85 |
View Full Product Details