Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACBP5
Entrez Gene ID
832825
Gene Name
acyl-CoA binding protein 5
Gene Aliases
AT5G27630, AtACBP5, F15A18.90, F15A18_90, acyl-CoA binding protein 5
Chromosome Location
Chr.5: 9775854 - 9781002 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001344021.1 NP_001331944.1 4 - 5 617 71
NM_001344022.1 NP_001331946.1 4 - 5 617 71
NM_001344023.1 NP_001331945.1 4 - 5 448 71
NM_122645.5 NP_198115.2 4 - 5 474 71
GenBank mRNA AY093156.1 - 4 - 5 448 71
BT008399.1 - 4 - 5 370 71

View Full Product Details