Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NEK3
Entrez Gene ID
832912
Gene Name
NIMA-related kinase 3
Gene Aliases
AT5G28290, ATNEK3, NIMA-RELATED KINASE3, NIMA-related kinase 3, T8M17.60, T8M17_60
Chromosome Location
Chr.5: 10278615 - 10282824 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001344066.1 NP_001330383.1 7 - 8 1170 99
NM_001344067.1 NP_001330382.1 8 - 9 687 99
NM_122712.3 NP_198181.1 8 - 9 897 99
GenBank mRNA AY090360.1 - 8 - 9 686 99
BT015415.1 - 7 - 8 434 99

View Full Product Details