Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MIS12
- Entrez Gene ID
-
833516
- Gene Name
- minichromosome instability 12 (mis12)-like protein
- Gene Aliases
-
ARABIDOPSIS MINICHROMOSOME INSTABILITY 12 (MIS12)-LIKE, AT5G35520, ATMIS12, MINICHROMOSOME INSTABILITY 12 (MIS12)-LIKE, MINICHROMOSOME INSTABILITY 12 HOMOLOGUE, MIS12 HOMOLOGUE, MOK9.12, MOK9_12
- Chromosome Location
-
Chr.5: 13701290 - 13703241 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_122943.3 |
NP_198402.2 |
3 - 4 |
418 |
68 |
View Full Product Details