Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
iqd33
Entrez Gene ID
833540
Gene Name
IQ-domain 33
Gene Aliases
AT5G35670, IQ-domain 33, MXH1.1, MXH1_1
Chromosome Location
Chr.5: 13855736 - 13858059 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_122958.4 NP_568529.1 4 - 5 1283 83
GenBank mRNA AK175704.1 - 4 - 5 914 83
AY088904.1 - 4 - 5 904 83

View Full Product Details