Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PHYC
Entrez Gene ID
833570
Gene Name
phytochrome C
Gene Aliases
AT5G35840, MIK22.15, MIK22_15, PHYTOCHROME C, phytochrome C
Chromosome Location
Chr.5: 14007812 - 14011848 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_122975.3 NP_198433.1 2 - 3 3077 62
GenBank mRNA X17343.1 - - 3058 62

View Full Product Details