Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
YAK1
Entrez Gene ID
833590
Gene Name
YAK1-like protein 1
Gene Aliases
AT5G35980, MEE13.9, MEE13_9, yeast YAK1-related gene 1
Chromosome Location
Chr.5: 14127960 - 14136351 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036893.2 NP_001031970.1 6 - 7 1339 99
NM_001344125.1 NP_001332507.1 6 - 7 817 99
NM_122989.4 NP_198447.2 6 - 7 1363 99
GenBank mRNA AK230350.1 - 6 - 7 1281 99
AY093090.1 - 6 - 7 1339 99
BT010549.1 - 6 - 7 772 99

View Full Product Details