Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
APE1
Entrez Gene ID
833856
Gene Name
acclimation of photosynthesis to environment
Gene Aliases
ACCLIMATION OF PHOTOSYNTHESIS TO ENVIRONMENT, ACCLIMATION OF PHOTOSYNTHESIS TO ENVIRONMENT, AT5G38660, MBB18.21, MBB18_21
Chromosome Location
Chr.5: 15470900 - 15476553 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203506.1 NP_001190435.1 5 - 6 688 111
NM_123227.6 NP_198682.3 5 - 6 1656 111
GenBank mRNA BT024514.1 - 5 - 6 600 111

View Full Product Details