Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABCB27
Entrez Gene ID
833896
Gene Name
transporter associated with antigen processing protein 2
Gene Aliases
ALS1, ALUMINUM SENSITIVE 1, ARABIDOPSIS THALIANA TRANSPORTER ASSOCIATED WITH ANTIGEN PROCESSING PROTEIN 2, AT5G39040, ATP-binding cassette B27, ATTAP2, AtALS1, MXF12.50, MXF12_50, TAP2, transporter associated with antigen processing protein 2
Chromosome Location
Chr.5: 15625460 - 15629835 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001344283.1 NP_001332027.1 10 - 11 1178 91
NM_123266.8 NP_198720.2 10 - 11 1242 91
GenBank mRNA AK229810.1 - 10 - 11 1124 91
DQ469798.1 - 10 - 11 1042 91

View Full Product Details