Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DEG9
Entrez Gene ID
834018
Gene Name
DegP protease 9
Gene Aliases
AT5G40200, DegP protease 9, DegP9, MSN9.100, MSN9_100, degradation of periplasmic proteins 9
Chromosome Location
Chr.5: 16070146 - 16073387 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_123384.3 NP_568577.1 1 - 2 863 69
GenBank mRNA AY046023.1 - 1 - 2 679 69
AY142608.1 - 1 - 2 607 69

View Full Product Details