Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MKK3
Entrez Gene ID
834042
Gene Name
mitogen-activated protein kinase kinase 3
Gene Aliases
AT5G40440, ATMKK3, MPO12.150, MPO12_150, mitogen-activated protein kinase kinase 3
Chromosome Location
Chr.5: 16181628 - 16184827 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001344328.1 NP_001318713.1 5 - 6 879 66
NM_001344329.1 NP_001332251.1 4 - 5 966 66
NM_001344330.1 NP_001332252.1 4 - 5 962 66
NM_001344331.1 NP_001332249.1 5 - 6 790 66
NM_001344332.1 NP_001332250.1 3 - 4 696 66
NM_123408.3 NP_198860.1 5 - 6 875 66
GenBank mRNA AB015314.1 - 5 - 6 741 66
AK117136.1 - 5 - 6 825 66
BT006481.1 - 4 - 5 711 66

View Full Product Details