Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATR
Entrez Gene ID
834082
Gene Name
Ataxia telangiectasia-mutated and RAD3-like protein
Gene Aliases
ARABIDOPSIS THALIANA ATAXIA TELANGIECTASIA-MUTATED AND RAD3-RELATED, AT5G40820, ATATR, ATRAD3, Ataxia telangiectasia-mutated and RAD3-related, MHK7.5, MHK7_5
Chromosome Location
Chr.5: 16342728 - 16353898 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001344355.1 NP_001332644.1 7 - 8 1625 130
NM_001344356.1 NP_001332643.1 7 - 8 1625 130
NM_001344357.1 NP_001332642.1 7 - 8 1625 130
NM_123447.3 NP_198898.2 7 - 8 1625 130

View Full Product Details