Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPP
Entrez Gene ID
834245
Gene Name
Insulinase (Peptidase family M16) family protein
Gene Aliases
AT5G42390, MDH9.8, MDH9_8, stromal processing peptidase
Chromosome Location
Chr.5: 16945165 - 16953353 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001344443.1 NP_001331624.1 15 - 16 2753 64
NM_123604.3 NP_199054.1 15 - 16 2756 64
GenBank mRNA BT000785.1 - 15 - 16 2670 64

View Full Product Details