Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SDG25
Entrez Gene ID
834246
Gene Name
SET domain protein 25
Gene Aliases
ARABIDOPSIS TRITHORAX-RELATED7, AT5G42400, ATXR7, MDH9.9, MDH9_9, SET domain protein 25
Chromosome Location
Chr.5: 16953515 - 16961245 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001344444.1 NP_001318731.1 2 - 3 375 113
NM_001344445.1 NP_001330664.1 2 - 3 377 113
NM_001344446.1 NP_001330665.1 2 - 3 366 113
NM_001344447.1 NP_001330668.1 2 - 3 374 113
NM_001344448.1 NP_001330669.1 2 - 3 374 113
NM_001344449.1 NP_001330666.1 2 - 3 375 113
NM_001344450.1 NP_001330667.1 1 - 2 197 113
NM_001344451.1 NP_001330670.1 2 - 3 373 113
GenBank mRNA EU014691.1 - 1 - 2 166 113

View Full Product Details