Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SDG25
Entrez Gene ID
834246
Gene Name
SET domain protein 25
Gene Aliases
ARABIDOPSIS TRITHORAX-RELATED7, AT5G42400, ATXR7, MDH9.9, MDH9_9, SET domain protein 25
Chromosome Location
Chr.5: 16953515 - 16961245 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001344444.1 NP_001318731.1 6 - 7 1751 127
NM_001344445.1 NP_001330664.1 6 - 7 1753 127
NM_001344446.1 NP_001330665.1 6 - 7 1742 127
NM_001344447.1 NP_001330668.1 6 - 7 1750 127
NM_001344448.1 NP_001330669.1 6 - 7 1750 127
NM_001344449.1 NP_001330666.1 6 - 7 1751 127
NM_001344450.1 NP_001330667.1 5 - 6 1573 127
NM_001344451.1 NP_001330670.1 6 - 7 1749 127
GenBank mRNA EU014691.1 - 5 - 6 1542 127

View Full Product Details