Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATS
Entrez Gene ID
834270
Gene Name
Homeodomain-like superfamily protein
Gene Aliases
ABERRANT TESTA SHAPE, AT5G42630, KAN4, KANADI 4, MFO20.5, MFO20_5
Chromosome Location
Chr.5: 17073673 - 17075904 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001125891.2 NP_001119363.1 1 - 2 502 72
NM_123627.3 NP_199077.1 1 - 2 550 72
GenBank mRNA AY048691.1 - 1 - 2 401 72
BT026053.1 - 1 - 2 393 72

View Full Product Details