Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRX3
Entrez Gene ID
834313
Gene Name
thioredoxin 3
Gene Aliases
AT5G42980, ATH3, ATTRX3, ATTRXH3, MBD2.18, MBD2_18, THIOREDOXIN, THIOREDOXIN H3, TRXH3, thioredoxin 3, thioredoxin H-type 3
Chromosome Location
Chr.5: 17242607 - 17244231 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_123664.4 NP_199112.1 2 - 3 378 75
GenBank mRNA AY059870.1 - 2 - 3 259 75
AY065098.1 - 2 - 3 259 75
AY085117.1 - 2 - 3 348 75
AY093318.1 - 2 - 3 213 75
AY114566.1 - 2 - 3 213 75
Z35474.1 - - 217 75

View Full Product Details