Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DCI1
Entrez Gene ID
834346
Gene Name
delta(3,5),delta(2,4)-dienoyl-CoA isomerase 1
Gene Aliases
''delta(3, 4)-DIENOYL-, 4)-DIENOYL-COA ISOMERASE 1, 4)-dienoyl-CoA isomerase 1, 4)-dienoyl-CoA isomerase 1'', 5), AT5G43280, ATDCI1, DELTA(2, DELTA(3, MNL12.10, MNL12_10, delta(2, delta(3
Chromosome Location
Chr.5: 17367827 - 17369510 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_123694.4 NP_199142.1 1 - 2 766 68
GenBank mRNA AY072351.1 - 1 - 2 705 68
BT002198.1 - 1 - 2 646 68

View Full Product Details