Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UAH
Entrez Gene ID
834380
Gene Name
ureidoglycolate amidohydrolase
Gene Aliases
ARABIDOPSIS THALIANA ALLANTOATE AMIDOHYDROLASE 2, AT5G43600, ATAAH-2, K9D7.10, K9D7_10, ureidoglycolate amidohydrolase
Chromosome Location
Chr.5: 17512440 - 17515513 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_123726.4 NP_199173.2 7 - 8 1110 72
GenBank mRNA AK230338.1 - - 2076 72
AY074343.1 - 7 - 8 1074 72
AY091387.1 - 7 - 8 899 72

View Full Product Details