Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TZP
Entrez Gene ID
834383
Gene Name
zinc knuckle (CCHC-type) family protein
Gene Aliases
AT5G43630, K9D7.13, K9D7_13
Chromosome Location
Chr.5: 17526660 - 17530605 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001344491.1 NP_001330182.1 4 - 5 2856 72
NM_001344492.1 NP_001330183.1 5 - 6 2685 72
NM_001344493.1 NP_001330181.1 5 - 6 2391 72
NM_123729.3 NP_199176.1 5 - 6 2406 72
GenBank mRNA BT003987.1 - 5 - 6 2384 72
BT005699.1 - 4 - 5 2255 72

View Full Product Details