Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HOT5
Entrez Gene ID
834417
Gene Name
GroES-like zinc-binding dehydrogenase family protein
Gene Aliases
ADH2, ALCOHOL DEHYDROGENASE 2, AT5G43940, ATGSNOR1, FALDH, GLUTATHIONE-DEPENDENT FORMALDEHYDE DEHYDROGENASE, GSNOR, MRH10.4, MRH10_4, PAR2, PARAQUAT RESISTANT 2, S-NITROSOGLUTATHIONE REDUCTASE, sensitive to hot temperatures 5
Chromosome Location
Chr.5: 17684156 - 17686534 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_123761.4 NP_199207.1 1 - 2 109 59
GenBank mRNA AK226412.1 - 1 - 2 100 59
AY039601.1 - 1 - 2 85 59
AY087250.1 - 1 - 2 97 59
BT010169.1 - 1 - 2 34 59

View Full Product Details