Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PHB7
Entrez Gene ID
834437
Gene Name
prohibitin 7
Gene Aliases
AT5G44140, ATPHB7, MLN1.6, MLN1_6, prohibitin 7
Chromosome Location
Chr.5: 17762491 - 17763666 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_123781.2 NP_199227.1 2 - 3 288 82

View Full Product Details