Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PP2-A6
Entrez Gene ID
834539
Gene Name
phloem protein 2-A6
Gene Aliases
AT5G45080, AtPP2-A6, K17O22.4, K17O22_4, phloem protein 2-A6
Chromosome Location
Chr.5: 18191355 - 18193328 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_123876.2 NP_199321.1 1 - 2 608 129

View Full Product Details