Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NRPC2
Entrez Gene ID
834550
Gene Name
nuclear RNA polymerase C2
Gene Aliases
AT5G45140, K18C1.1, K18C1_1, nuclear RNA polymerase C2
Chromosome Location
Chr.5: 18247149 - 18258197 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001344590.1 NP_001332060.1 14 - 15 1243 100
NM_001344591.1 NP_001332059.1 14 - 15 1161 100
NM_123882.5 NP_199327.4 14 - 15 1161 100

View Full Product Details