Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BMY2
Entrez Gene ID
834566
Gene Name
beta-amylase 2
Gene Aliases
AT5G45300, BAM8, BETA-AMYLASE 8, K9E15.8, K9E15_8, beta-amylase 2
Chromosome Location
Chr.5: 18353525 - 18356888 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036937.1 NP_001032014.1 7 - 8 1632 91
NM_001344606.1 NP_001330613.1 7 - 8 1721 91
NM_123898.3 NP_199343.1 7 - 8 1636 91
GenBank mRNA AK117140.1 - 7 - 8 1633 91
BT006482.1 - 7 - 8 1610 91

View Full Product Details