Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MEE62
Entrez Gene ID
834620
Gene Name
Leucine-rich repeat protein kinase family protein
Gene Aliases
AT5G45800, MRA19.24, MRA19_24, maternal effect embryo arrest 62
Chromosome Location
Chr.5: 18575520 - 18579241 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_123948.5 NP_199392.1 1 - 2 1650 125
GenBank mRNA AY072097.1 - 1 - 2 1506 125
AY096584.1 - 1 - 2 1381 125
FJ708790.1 - 1 - 2 1381 125

View Full Product Details