Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FLS2
Entrez Gene ID
834676
Gene Name
Leucine-rich receptor-like protein kinase family protein
Gene Aliases
AT5G46330, FLAGELLIN-SENSITIVE 2, MPL12.13, MPL12.8, MPL12_13
Chromosome Location
Chr.5: 18791575 - 18795908 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001344672.1 NP_001330009.1 1 - 2 3364 69
NM_124003.4 NP_199445.1 1 - 2 3364 69
GenBank mRNA FJ708792.1 - 1 - 2 3137 69

View Full Product Details