Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRP39-2
Entrez Gene ID
834683
Gene Name
Tetratricopeptide repeat (TPR)-like superfamily protein
Gene Aliases
AT PRP39-2, AT5G46400, MPL12.20, MPL12_20
Chromosome Location
Chr.5: 18819933 - 18824782 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001344676.1 NP_001330626.1 2 - 3 426 124
NM_124010.3 NP_199452.2 4 - 5 685 124

View Full Product Details