Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AGD9
Entrez Gene ID
834718
Gene Name
ARF-GAP domain 9
Gene Aliases
AGD8, ARF-GAP domain 8, ARF-GAP domain 9, AT5G46750, MZA15.17, MZA15_17
Chromosome Location
Chr.5: 18969736 - 18971981 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_124045.5 NP_199487.1 5 - 6 1161 89
GenBank mRNA AY099693.1 - 5 - 6 1085 89
AY128872.1 - 5 - 6 997 89

View Full Product Details