Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LBA1
Entrez Gene ID
834747
Gene Name
RNA helicase
Gene Aliases
AT5G47010, ATUPF1, LOW-LEVEL BETA-AMYLASE 1, MQD22.15, MQD22_15, UPF1
Chromosome Location
Chr.5: 19071824 - 19079453 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_124072.3 NP_199512.2 21 - 22 2924 96

View Full Product Details