Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- NUDT8
- Entrez Gene ID
-
834771
- Gene Name
- nudix hydrolase homolog 8
- Gene Aliases
-
AT5G47240, MQL5.10, MQL5_10, atnudt8, nudix hydrolase homolog 8
- Chromosome Location
-
Chr.5: 19183523 - 19185644 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_124095.4 |
NP_568680.1 |
6 - 7 |
1119 |
67 |
View Full Product Details