Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FPS1
Entrez Gene ID
834828
Gene Name
farnesyl diphosphate synthase 1
Gene Aliases
AT5G47770, FARNESYL DIPHOSPHATE SYNTHASE 1, MCA23.9, MCA23_9, farnesyl diphosphate synthase 1
Chromosome Location
Chr.5: 19345110 - 19347677 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_124151.3 NP_199588.1 6 - 7 859 97
GenBank mRNA AF370324.1 - 6 - 7 796 97
AY063112.1 - 6 - 7 672 97
U80605.1 - 6 - 7 699 97

View Full Product Details