Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GAUT4
Entrez Gene ID
834829
Gene Name
galacturonosyltransferase 4
Gene Aliases
AT5G47780, JS36L, LGT3, MCA23.10, MCA23_10, galacturonosyltransferase 4
Chromosome Location
Chr.5: 19347874 - 19350976 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_124152.3 NP_568688.1 5 - 6 480 74
GenBank mRNA AY056202.1 - 5 - 6 428 74
AY091394.1 - 5 - 6 363 74
KJ138647.1 - 5 - 6 363 74

View Full Product Details