Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AMK2
Entrez Gene ID
834835
Gene Name
adenosine monophosphate kinase
Gene Aliases
AT5G47840, MCA23.18, MCA23_18, adenosine monophosphate kinase
Chromosome Location
Chr.5: 19375317 - 19378465 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001344749.1 NP_001330051.1 1 - 2 264 122
NM_124158.4 NP_199595.1 1 - 2 335 122
GenBank mRNA AY049305.1 - 1 - 2 206 122
BT001036.1 - 1 - 2 164 122

View Full Product Details