Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLD5
Entrez Gene ID
834960
Gene Name
DNA replication protein-like protein
Gene Aliases
AT5G49010, EMB2812, EMBRYO DEFECTIVE 2812, K19E20.14, K19E20_14, SYNTHETIC LETHALITY WITH DPB11-1 5
Chromosome Location
Chr.5: 19866231 - 19868877 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001125927.2 NP_001119399.1 3 - 4 605 71
NM_124278.4 NP_199712.2 3 - 4 605 71
GenBank mRNA AK221044.1 - 3 - 4 605 71
BT010406.1 - 3 - 4 372 71

View Full Product Details