Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- APRR4
- Entrez Gene ID
-
834984
- Gene Name
- pseudo-response regulator 4
- Gene Aliases
-
AT5G49240, K21P3.12, K21P3_12, PRR4, PSEUDO-RESPONSE REGULATOR 4, pseudo-response regulator 4
- Chromosome Location
-
Chr.5: 19962934 - 19964351 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_124301.1 |
NP_199735.1 |
3 - 4 |
743 |
92 |
View Full Product Details