Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GH9A1
Entrez Gene ID
835035
Gene Name
glycosyl hydrolase 9A1
Gene Aliases
AT5G49720, ATGH9A1, CELLULASE, DEC, DEFECTIVE CYTOKINESIS, IRREGULAR XYLEM 2, IRX2, K2I5.8, K2I5_8, KOR, KOR1, KORRIGAN, KORRIGAN 1, RADIALLY SWOLLEN 2, RSW2, TSD1, TUMOROUS SHOOT DEVELOPMENT 1, glycosyl hydrolase 9A1
Chromosome Location
Chr.5: 20197391 - 20200468 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_124350.3 NP_199783.1 5 - 6 1750 122
GenBank mRNA AF074375.1 - 5 - 6 1513 122
U37702.1 - 5 - 6 1520 122

View Full Product Details