Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MMT
Entrez Gene ID
835044
Gene Name
methionine S-methyltransferase
Gene Aliases
AT5G49810, methionine S-methyltransferase
Chromosome Location
Chr.5: 20239293 - 20246398 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_124359.4 NP_199792.1 3 - 4 750 160
GenBank mRNA AY094459.1 - 3 - 4 700 160
BT002664.1 - 3 - 4 625 160

View Full Product Details